missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]pyridin-2-ylmethanol, 97%, Thermo Scientific™

Catalog Number p-3176170
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
131337-81-0
C8H7NOS
165.21
MFCD07772792
XLUKSWZEEAGBDH-UHFFFAOYSA-N
thieno 2,3-b pyridin-2-ylmethanol, thieno 2,3-b pyridine-2-methanol, thieno 2,3-b pyridin-2-yl methanol, acmc-1bz5x, thieno 2,3-b pyridine-2-ylmethanol, 2-hydroxymethylthieno 2,3-b pyridine, thiopheno 2,3-b pyridin-2-ylmethan-1-ol
7162092
thieno[2,3-b]pyridin-2-ylmethanol
C1=CC2=C(N=C1)SC(=C2)CO
This item is not returnable. View return policy
131337-81-0
C8H7NOS
165.21
MFCD07772792
XLUKSWZEEAGBDH-UHFFFAOYSA-N
thieno 2,3-b pyridin-2-ylmethanol, thieno 2,3-b pyridine-2-methanol, thieno 2,3-b pyridin-2-yl methanol, acmc-1bz5x, thieno 2,3-b pyridine-2-ylmethanol, 2-hydroxymethylthieno 2,3-b pyridine, thiopheno 2,3-b pyridin-2-ylmethan-1-ol
7162092
thieno[2,3-b]pyridin-2-ylmethanol
C1=CC2=C(N=C1)SC(=C2)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 165.22
Percent Purity 97%
Chemical Name or Material Thieno[2, 3-b]pyridin-2-ylmethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'