missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine, 98%, Thermo Scientific Chemicals

Catalog Number p-2692307
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
504-78-9
C3H8NS
90.16
MFCD00005211
OGYGFUAIIOPWQD-UHFFFAOYSA-O
thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
10444
CHEBI:50120
C1CSC[NH2+]1
This item is not returnable. View return policy
504-78-9
C3H8NS
90.16
MFCD00005211
OGYGFUAIIOPWQD-UHFFFAOYSA-O
thiazolidine, tetrahydrothiazole, 1-thia-3-azacyclopentane, thiazole, tetrahydro, unii-i320806bkw, ccris 4275, acmc-1ao0m, 4-27-00-00009 beilstein handbook reference
10444
CHEBI:50120
C1CSC[NH2+]1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1310g/mL
Boiling Point 72°C to 75°C (25.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5525 to 1.5545
Quantity 1 g
Specific Gravity 1.131
IUPAC Name 1,3-thiazolidin-3-ium
Formula Weight 89.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Thiazolidine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thiazolidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'