missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thiazolidine-2,4-dicarboxylic acid, 97%

Brand:  Alfa Aesar™ H62405.03

Code : NEW

Additional Details : CAS Number : 30097-06-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498908

  • £81.94 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Thiazolidine-2,4-dicarboxylic acid
30097-06-4
177.18
MFCD00145399

Specifications

Thiazolidine-2,4-dicarboxylic acid
1g
Solid
177.18
White
30097-06-4
97%
MFCD00145399
Videos
SDS