missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazole-5-methanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Density | 1.3200g/mL |
Assay Percent Range | 94% min. (GC) |
Quantity | 25 g |
Specific Gravity | 1.32 |
Formula Weight | 115.15 |
Percent Purity | 95% |
Chemical Name or Material | Thiazole-5-methanol |
Safety and Handling
Product Identifier
- Thiazole-5-methanol
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- H412-Harmful to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'