missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazole-2-carboxaldehyde, 95%, Thermo Scientific Chemicals
22.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.
Solubility
Fully miscible with water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.
Solubility
Fully miscible with water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Color | Colorless to Yellow |
Density | 1.288 g/mL |
Boiling Point | 61°C to 63°C (15 mmHg) |
Flash Point | 68°C (154°F) |
Refractive Index | 1.574 |
Quantity | 1 g |
Solubility Information | Fully miscible with water. |
Formula Weight | 113.13 |
Percent Purity | 95% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'