missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 100 g |
Formula Weight | 337.27 |
Grade | FCC/USP |
Chemical Name or Material | Thiamine Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'