Learn More
Nickel Electro™ Thermometer Clip
Nickel Electro™ Thermometer Clip reduces the risk of breakage and comes complete with bent stem thermometer to fit onto water baths.
Brand:  Nickel Electro™ TC-1
Code : 03
Additional Details : Weight : 0.10000kg
Product Code. 11409539
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Thermometer fitted with two rubber O rings ensuring a snug fit inside the clip
  • Reduces the chances of breakages
  • Temperatures can be easily read even when a SL range accessory lid is fitted
  • Sticky pads are fitted to the body of the clip preventing it from scratching the water bath
  • Used as a reference to measure actual water temperature of the water bath
Specifications
Specifications
Thermometer Clip
Stainless Steel
15 x 1 cm
Bent Stem Spirit Filled Thermometer