Learn More
Thermo Scientific™ VersiPlate PCR Strip Tube Plate, 96-well, low profile, white
Flexible 96-well plates of low profile strip tubes for use in PCR and qPCR applications.
Brand: Thermo Scientific™ AB1800W
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific VersiPlates are unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
The Thermo Scientific VersiPlates are unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Highlights
- Easier sample handling as multiple strips of samples can be handled simultaneously
- Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
- Can be used in instruments with or without a VersiFrame.
- The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
- The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
- Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
- Maximum fill volume of 0.2 mL
- Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals
- PCR
- qPCR/Real-Time PCR
Certifications
Certified free from human DNA, RNases, and DNases
Specifications
No Barcode | |
25 Plate | |
White | |
Real-Time PCR Systems or Thermal Cyclers | |
0.2 mL | |
0.2 mL |
Certified RNase/DNase-free | |
VersiPlate | |
PCR Microplate | |
Polycarbonate | |
96 | |
Low profile |