missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TraceGOLD™ TG-WaxMS GC Columns

Manufactured for better column-to-column reproducibility

Brand:  Thermo Scientific™ 26088-2760

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 13438018

  • £358.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Polar phase, polyethylene glycol
  • Polar-deactivated surface tightly binds polymer for excellent thermal stability
  • Resist oxidative damage, damage from strongly acidic or basic volatiles better than silicone solid phases
  • Equivalent to USP G14, G15, G16, G20 and G39 phases
  • Similar to DB-WAX, DB-WAXetr, HP-Wax, HP-Innowax, Supelcowax 10, CP-Wax 52 CB, Stabilwax, Rtx-Wax, BP20, ZB-Wax, Optima Wax, AT-Wax

Specifications

Specifications

20m
40 to 250/260°C
Polyethylene Glycol
GC Capillary Column
0.15μm
0.15mm
Videos