missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Legend™ T3/RT3 Swinging Bucket Rotor

Versatile high-throughput rotor for use with Legend™ T3 and RT3 centrifuges

Brand:  Thermo Scientific™ 11210823

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11774928

  • £2556.80 / 4 sets


Call for latest availability
+44 (0)1509 555 500
Add to basket

Requires : Buckets or carriers (order separately)

Description

Description

Accepts buckets or carriers to spin tubes, bottles, microplates and special containers.

  • Capacity: 4 × 750mL
  • Designed to be loaded into centrifuge with buckets/carriers already in place
  • Sealed buckets 05-376-262 offer biohazard protection
  • Transparent sealing caps allow visual detection of biological risks

Specifications

Specifications

Unsealed Plate Carrier
3,094 x g
M4 High Throughput swing Rotor
4 x 750mL
4,000rpm
Rotor body only
Videos