missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Shandon™ Pathcentre™ Tissue Processor Accessories
For use with the Thermo Scientific Shandon Tissue Processor
Brand: Thermo Scientific™ 66110048
Code : 03
Additional Details : Weight : 1.00000kg
Specifications
Airbag | |
Tissue processor | |
4 Pack |