Learn More
Thermo Scientific™ PocketTips™
Nanoliter dispensing pipette tips with an internally molded capillary pocket that holds 50 to 500nL
Brand:  Thermo Scientific™ U200-250
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12927227
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
A broad range of pocket volumes are compatible with many industry standard 96- and 384-channel automated liquid handling platforms to allow existing equipment to implement new innovative pipetting procedures.
  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing—thorough mixing without damaging sensitive material
  • Consistent accuracy and precision; coefficient of variation of less than 6% can be achieved for any pocket size
  • Color: Clear
Specifications
Specifications
PerkinElmer™Ć¾Hamilton™ MICROLAB™ STAR
Automation
250nL
PerkinElmer™, Hamilton™ MICROLAB™ STAR
Videos