Learn More
Invitrogen™ Human TIMP1 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP77537
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
TIMP1 Recombinant Protein, RP-77537, from Invitrogen.
TIMP1 (metalloproteinase inhibitor 1) metalloproteinase inhibitor that functions by forming one to one complexes with target metalloproteinases, such as collagenases, and irreversibly inactivates them by binding to their catalytic zinc cofactor. Acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP13 and MMP16. Does not act on MMP14. Also functions as a growth factor that regulates cell differentiation, migration and cell death and activates cellular signaling cascades via CD63 and ITGB1. RP-77537 is purified from human neutrophil granulocytes. RP-77537 may be used as a control with corresponding anti-TIMP 1 antibodies.Specifications
P01033 | |
TBS with 0.05% Brij-35, 1µM ZnCl2, 5mM CaCl2 and 0.05% sodium azide; pH 7 | |
Human TIMP1 | |
10μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
TIMP 1 | |
TIMP1 | |
Unconjugated | |
Baculovirus | |
Liquid | |
purified |
Control | |
7076 | |
7 | |
Insect Cells | |
RUO | |
Human TIMP1 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only.