missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Yeast Thioredoxin 2, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ LFP0052
Additional Details : Weight : 0.99000kg
Description
Thioredoxin 2 (Ys) Recombinant Protein, LF-P0052, from Invitrogen.
Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cysteine residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds. Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations. Specific protein disulfide targets for reduction by Trx include protein disulfide isomerase (PDI) and a number of transcription factors such as p53 , NF-kB and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins. This protein corresponds to yeast Thioredoxin 2 (1-104 aa), which has a molecular weight of 11.3 kDa. Specific activity: 9.7 U/mg, where one unit will cause a delta-A650 of 1 in 1 min at 25°C in the insulin reduction assay. Purity: >95% by SDS-PAGE.Specifications
1mg/mL | |
0.02M HEPES with 10% glycerol and no preservative; pH 7.0 | |
7.0 | |
E. coli | |
RUO | |
TRX2 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control | |
Yeast Thioredoxin 2 | |
0.5mg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Yeast Thioredoxin 2 | |
Yeast | |
Ys | |
Recombinant | |
Recombinant |
in vitro research only.