missing translation for 'onlineSavingsMsg'
Learn More

RuvC Recombinant Protein, Invitrogen™

RuvC Recombinant Protein, RP-4911, from Invitrogen™

Brand:  Invitrogen RP4911

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11564191

  • £1928.39 / 100µg


Estimated Shipment
15-02-2018
Add to basket

Description

Description

E. coli RuvC protein is a structurally specific endonuclease which binds specifically to the Holliday structure, an intermediate of recombination, at the late stage of homologous recombination and recombination repair and introduces a nick in the symmetrical point of the Holliday junction cleaving and resolving the recombinant. The product is a recombinant protein abundantly expressed by E. coli and purified by methods such as chromatography. Its molecular weight is 19 kDa and forms a dimer in liquid and in binding state with the Holliday structure.
Host Species: E. Coli
Formulation: 100μg purified protein (13mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RP-4911 can be used as a positive control in Western blotting, as well as in studies in studies on homologous recombination mechanism. RP-4911 calculated molecular weight is ∽19 kDa. Purity is 90%.
Specifications

Specifications

Control, Western Blot
100μg purified protein (13mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RuvC Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Purified
∽19
7.5
100μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Videos