missing translation for 'onlineSavingsMsg'
Learn More

RuvB Recombinant Protein, Invitrogen™

RuvB Recombinant Protein, RP-4908, from Invitrogen™

Brand:  Invitrogen RP4908

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11534191

  • £760.70 / 20µg


Estimated Shipment
22-06-2018
Add to basket

Description

Description

E. coli RuvB protein forms a complex with RuvA protein at the late stage of homologous recombination and recombination repair and binds specifically to the Holliday structure which is the intermediate of recombination, allowing the migration of Holliday junction using ATP hydrolysis energy and expands the heteroduplex region. RuvB forms a hexamer ring structure and surrounds the double chain DNA and covers RuvA tetramer bound to the Holliday junction from both sides. RuvB is a DNA motor protein which possesses the ATPase activity, activated by DNA and RuvA protein. Its molecular weight is 37 kD and forms a dimer in solution in the physiological condition.
Host Species: E. Coli
Formulation: 20μg purified protein (2mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RP-4908 can be used as a positive control in Western blotting, as well as in studies on homologous recombination mechanism. RP-4908 calculated molecular weight is ∽37 kDa. Purity is 90%.
Specifications

Specifications

Control, Western Blot
20μg purified protein (2mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RuvB Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Purified
∽37
7.5
20μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Videos