missing translation for 'onlineSavingsMsg'
Learn More

RuvA Recombinant Protein, Invitrogen™

RuvA Recombinant Protein, RP-4907, from Invitrogen™

Brand:  Invitrogen RP4907

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11524191

  • £1928.39 / 100µg


Estimated Shipment
15-02-2018
Add to basket

Description

Description

E. coli RuvA protein binds specifically to the Holliday structure which is the intermediate of recombination at the late stage of homologous recombination and recombination repair and forms a complex with RuvB motor protein allowing the migration of Holliday junction using ATP hydrolysis energy and expands the heteroduplex region. In solution, it forms a tetramer and binds to the cross-like DNA of the Holliday junction from below and above holding it in between.
Host Species: E. Coli
Formulation: 100μg purified protein (2.7mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RP-4907 can be used as a positive control in Western blotting, as well as in studies on homologous recombination mechanism. RP-4907 calculated molecular weight is ∽22 kDa. Purity is 90%.
Specifications

Specifications

2.7mg/mL
Purified
∽22
7.5
100μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Control, Western Blot
100μg purified protein (2.7mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RuvA Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Videos