missing translation for 'onlineSavingsMsg'
Learn More

RuvA Recombinant Protein, Invitrogen™

RuvA Recombinant Protein, RP-4906, from Invitrogen™

Brand:  Invitrogen RP4906

UNSPSC : 12352203

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11514191

  • £638.00 / 20µg


Estimated Shipment
21-08-2017
Add to basket

Specifications

Description

E. coli RuvA protein binds specifically to the Holliday structure which is the intermediate of recombination at the late stage of homologous recombination and recombination repair and forms a complex with RuvB motor protein allowing the migration of Holliday junction using ATP hydrolysis energy and expands the heteroduplex region. In solution, it forms a tetramer and binds to the cross-like DNA of the Holliday junction from below and above holding it in between.
Host Species: E. Coli
Formulation: 20μg purified protein (2.7mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RP-4906 can be used as a positive control in Western blotting, as well as in studies on homologous recombination mechanism. RP-4906 calculated molecular weight is ∽22 kDa. Purity is 90%.
Description & Specifications

Specifications

2.7mg/mL
Purified
∽22
7.5
20μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Control, Western Blot
20μg purified protein (2.7mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RuvA Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes