missing translation for 'onlineSavingsMsg'
Learn More

IL-2 (human) Recombinant Protein, Invitrogen™

IL-2 (human) Recombinant Protein, RP-75587, from Invitrogen™

Brand:  Invitrogen RP75587

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11568911

  • £202.90 / 10µg


Estimated Shipment
18-05-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 10μg purified protein (0.44mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8
RP-75587 contains purified recombinant human IL-2 protein with an N-terminal hexahistidine tag. RP-75587 may be used as a positive control in many immunological assays. Purity >90% by SDS-PAGE.
Specifications

Specifications

0.05mg/mL
Purified
Human IL-2 Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Control, Western Blot
10μg purified protein (0.44mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8
10μg
N/A
Unlabeled
E. coli
Videos