missing translation for 'onlineSavingsMsg'
Learn More

IL-2 (human) Recombinant Protein, Invitrogen™

IL-2 (human) Recombinant Protein, RP-75589, from Invitrogen™

Brand:  Invitrogen RP75589

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11588911

  • £2153.84 / 1mg


Estimated Shipment
16-02-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75589 contains purified recombinant human IL-2 protein with an N-terminal hexahistidine tag. RP-75589 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
1mg purified protein in PBS with 50% glycerol and no preservative
1mg
N/A
Unlabeled
E. coli
Purified
Human IL-2 Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Videos