missing translation for 'onlineSavingsMsg'
Learn More

IL-2 (human) Recombinant Protein, Invitrogen™

IL-2 (human) Recombinant Protein, RP-75588, from Invitrogen™

Brand:  Invitrogen RP75588

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11578911

  • £443.32 / 100µg


Estimated Shipment
21-02-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
RP-75588 contains purified recombinant human IL-2 protein with an N-terminal hexahistidine tag. RP-75588 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
8
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Purified
Human IL-2 Recombinant Protein
100μg
N/A
Unlabeled
E. coli
Videos