missing translation for 'onlineSavingsMsg'
Learn More

IL-10 (human) Recombinant Protein, Invitrogen™

IL-10 (human) Recombinant Protein, RP-75612, from Invitrogen™

Brand:  Invitrogen RP75612

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11518941

  • £429.24 / 100µg


Estimated Shipment
23-03-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein (0.25mg/mL) in 0.01M tris HCl with 50% glycerol, 0.25M NaCl and no preservative; pH 8
RP-75612 contains purified recombinant human IL-10 protein with an N-terminal hexahistidine tag. RP-75612 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein (0.25mg/mL) in 0.01M tris HCl with 50% glycerol, 0.25M NaCl and no preservative; pH 8
8
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Purified
Human IL-10 Recombinant Protein
100μg
N/A
Unlabeled
E. coli
Videos