missing translation for 'onlineSavingsMsg'
Learn More

RAD52 (human) Recombinant Protein, Invitrogen™

RAD52 (human) Recombinant Protein, RP-4973, from Invitrogen™

Brand:  Invitrogen RP4973

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11524211

  • £1797.82 / 100µg


Estimated Shipment
20-03-2018
Add to basket

Description

Description

Human Rad52 protein plays a major role in genetic recombination and recombination repair by mediating strand-annealing reaction between complementary DNA strands. Rad52 functionally and physically interacts with Rad51, Dmc1, and RPA in recombination processes. In radiation damaged cell, Rad52 is phosphorylated at Tyr104 by c-Abl tyrosine kinase and forms nuclear foci.
Host Species: E. coli
Formulation: 100μg purified protein (1mg/mL) in 0.02M potassium phosphate with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; pH 7.5
RP-4973 contains highly purified recombinant Rad52 protein expressed in E. coli. RP-4973 may be used as a control in Western blotting applications.
Specifications

Specifications

1mg/mL
Purified
Human RAD52 Recombinant Protein
100μg
Store at -80°C, Avoid Freeze/Thaw Cycles
Human
Yes
Control, Western Blot
100μg purified protein (1mg/mL) in 0.02M potassium phosphate with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; pH 7.5
7.5
E. coli
Unlabeled
E. coli
Videos