missing translation for 'onlineSavingsMsg'
Learn More

RAD51 Recombinant Protein, Invitrogen™

RAD51 Recombinant Protein, RP-4964, from Invitrogen™

Brand:  Invitrogen RP4964

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11594201

  • £663.01 / 20µg


Estimated Shipment
26-06-2018
Add to basket

Description

Description

Human Rad51 protein is a functional and structural homolog of E. coli RecA protein, which plays a major role in genetic recombination and recombination repair by mediating strand exchange reaction between homologous DNA strands. Rad51 functionally and physically interacts with its paralogs Dmc1, Rad51B, Rad51C, Rad51D, Xrcc2 and Xrcc3, and also with Rad52 in recombination processes. It also interacts with oncogene proteins and tumor suppressors such as BRCA1, BRCA2, and P53 for the maintenance of genome stability.
Host Species: E. Coli
Formulation: 20μg purified protein (1mg/mL) in tris HCl with 0.01M KCl, 10% glycerol, 1mM DTT, 0.5mM EDTA and no preservative; pH 8
RP-4964 has been successfully used in Western blot proceedures. It is specific for the ∽35 kDa Rad51 protein. The Human gene Rad51 is homologous to the highly conserved bacterial gene RecA. RP-4964 is highly purified from E. coli over-expressing human Rad51 protein as a recombinant protein. The tag was removed from the recombinant protein (though it still contains Gly-Ser-His at the N-terminal). The protein has been shown to retain nuclear filament forming and strand-exchange activity as well as interaction with Rad52. RP-4964 was confirmed to possess single strand DNA stimulated ATPase activity.
Specifications

Specifications

1mg/mL
Purified
∽35
8
E. coli
RAD51 homolog
Human
Yes
Control, Western Blot
20μg purified protein (1mg/mL) in tris HCl with 0.01M KCl, 10% glycerol, 1mM DTT, 0.5mM EDTA and no preservative; pH 8
Human Rad51 Recombinant Protein
20μg
Store at -80°C, Avoid Freeze/Thaw Cycles
Unlabeled
E. coli
Videos