Learn More
Human IgG Isotype Control, Invitrogen™
Human Isotype Control
Brand: Invitrogen 31154
190.65 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : PN
Additional Details : Weight : 0.80000kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity/Antibody Specificity: The precipitin lines against goat anti-human whole serum are the same as those against goat anti-human IgG, Fc fragment specific when assayed by immunoelectrophoresis. No cross-reactivity was observed against goat anti-human IgM, Fc5-mu fragment specific or goat anti-human IgA, alpha chain specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Storage: After opening, dilute only enough antibody for a single day's use. The human source material used to make this antibody has been tested and found negative for Anti-HIV-1/HIV-2),HCV antibodies, HBsAg and ALT (SGPT). Country of Origin: USA
Human IgG Control antibody specifically detects Human IgG in Not Applicable samples. It is validated for Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry.Specifications
Human IgG | |
11.9 mg/mL | |
PBS with no preservative; pH 7.6 | |
Human | |
10 mg | |
Isotype Control (Primary) | |
4° C | |
IgG |
Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG | |
purified | |
RUO | |
Not Applicable | |
Liquid |