Thermo Scientific™ Pierce™ Primary Neuron Isolation Kit

Easily isolate and culture highly viable functional primary neurons

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 88280

UNSPSC: 41116012

Code: NEW

Additional Details:
Additional Details: Weight: 0.50000kg



Product Code. 15285713

Quantity Price
1 £ 379.0 / 1 set
EU Stock 1
Add to basket


Description and Specification

Specification

Sufficient For Isolation and culture of primary neurons from 50 pairs of embryonic mouse/rat cortexes or 150 pairs of embryonic mouse/rat hippocampi
Formulation Six-component kit
Includes Neuronal Culture Medium, 500mL Hanks' Balanced Salt Solution (HBSS without Ca2+, Mg2+), 500mL Neuronal Isolation Enzyme (with papain), 5 vials Neuronal Culture Medium Supplement (50X), 10mL Glutamine Supplement (100X), 5mL Neuronal Growth Supplement (1000X), 0.5mL
Quantity 1L kit
Product Type Primary Neuron Isolation Kit