Thermo Scientific™ Pierce™ Primary Cardiomyocyte Isolation Kit

Easily isolate and culture highly viable functional primary cardiomyocytes

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 88281

PIERCE PRIMARY CARDIOMYOCYTE ISOLATION KIT

UNSPSC: 41116012

Code: NEW

Additional Details:
Additional Details: Weight: 0.50000kg



Product Code. 15295713

Quantity Price
1 £ 368.0 / 1 set
EU Stock 12
Add to basket


Description and Specification

Specification

Quantity 1L kit
Sufficient For Isolation and culture of cardiomyocytes from 50 neonatal mouse/rat hearts
Formulation Five-component kit
Includes DMEM for Primary Cell Isolation, 500mL Hanks' Balanced Salt Solution (HBSS without Ca2+, Mg2+), 500mL Cardiomyocyte Isolation Enzyme 1 (with papain), lyophilized, 5 vials Cardiomyocyte Isolation Enzyme 2 (with thermolysin) (20X), 0.1mL, 5 vials Cardiomyocyte Growth Supplement (1000X), 0.5mL
Product Type Primary Cardiomyocyte Isolation Kit