Learn More
Thermo Scientific™ Pierce™ Premium Grade SPDP
Crosslink proteins and other biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade SPDP, a high-quality formulation of this popular pyridyldithiol-based amine-to-sulfhydryl crosslinker.
£445.00
Specifications
Product Line | Pierce™ |
---|---|
Labeling Method | Chemical Labeling |
Crosslinker Type | Heterobifunctional |
Reactive Moiety | NHS Ester, Pyridyldisulfide |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15372627
|
Thermo Scientific™
PG82087 |
100 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific Pierce Premium Grade SPDP is our highest quality formulation of this pyridyldithiol-based amine-to-sulfhydryl crosslinker, specially characterized for applications where product integrity and risk minimization are paramount. SPDP is a disulfide crosslinker used for reversible (reducible) amine-to-sulfhydryl conjugation.
- High quality - identity and purity confirmed by multiple tests, including quantitative NMR
- Product integrity - enhanced level of testing and characterization compared to standard grade
- Lot retention - ample supply of past lots retained to help ensure future process testing
- Change management - Change Control Notification (CCN) service
- Consistent manufacture - batch-specific manufacturing documentation review
SPDP is N-succinimidyl 3-(2-pyridyldithio)-propionate, a crosslinker for amine-to-sulfhydryl conjugation that forms cleavable (reducible) disulfide bonds with cysteine sulfhydryls. At one end is an N-hydroxysuccinimide (NHS) ester that will react with a primary amine to form a stable amide bond. At the other end is a pyridyl disulfide group that will react with a sulfhydryl group to form a reversible disulfide bond.
Specifications
Pierce™ | |
Heterobifunctional | |
Powder | |
Crosslinker |
Chemical Labeling | |
NHS Ester, Pyridyldisulfide | |
DMF, DMSO |