missing translation for 'onlineSavingsMsg'
Learn More

Rat AA-Nat (residues 24-33) Synthetic Peptide, Invitrogen™

Rat AA-Nat (residues 24-33) Synthetic Peptide, PEP-063, from Invitrogen™

Brand:  Invitrogen PEP063

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11544720

  • £100.16 / 50µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-063 is a 10 amino acid synthetic peptide whose sequence corresponds to residues 24-33 of rat AA-NAT. The sequence of this peptide is (amino to carboxy terminus): C24-Q-R-R-H-T-L-P-A-S33 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and AA-NAT, catalog # PA1-760. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.
Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
C24-Q-R-R-H-T-L-P-A-S33
Rat AA-Nat (Residues 24-33) Synthetic Peptide
Videos