missing translation for 'onlineSavingsMsg'
Learn More

Rat and Human SAP97 (residues 16-33) Synthetic Peptide, Invitrogen™

Rat and Human SAP97 (residues 16-33) Synthetic Peptide, PEP-022, from Invitrogen™

Brand:  Invitrogen PEP022

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11544680

  • £100.16 / 50µg


Estimated Shipment
13-03-2018
Add to basket

Description

Description


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-022 is an 18 amino acid (amino acids 16-33) synthetic peptide whose sequence is found in rat and human SAP97. The sequence of this peptide is (amino to carboxy terminus): E16-E-Y-R-S-K-L-S-Q-T-E-D-R-Q-L-R-S-S33 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SAP97, catalog # PA1-044. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 ml of distilled water.
Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
E16-E-Y-R-S-K-L-S-Q-T-E-D-R-Q-L-R-S-S33
Rat and Human SAP97 (Residues 16-33) Synthetic Peptide
Videos