missing translation for 'onlineSavingsMsg'
Learn More

Human IRS-1 (residues 301-315) Synthetic Peptide, Invitrogen™

Human IRS-1 (residues 301-315) Synthetic Peptide, PEP-191, from Invitrogen™

Brand:  Invitrogen PEP191

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11594840

  • £86.12 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-191 is a 15 amino acid synthetic peptide corresponding to amino acid residues 301-315 from human IRS-1. The sequence of this peptide is (amino to carboxy terminus): E S I T A T S P A S M V G G K (NH2). This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1057. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.
Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
E S I T A T S P A S M V G G K(NH2)
Human IRS-1 (Residues 301-315) Synthetic Peptide
Videos