Learn More
Invitrogen™ Human Estrogen Receptor beta (aa 467-485) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP029
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human Estrogen Receptor B (ERb) (resides 467-485) Synthetic Peptide, PEP-029, from Invitrogen.
Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease. PEP-029 is a 19 amino acid (amino acids 467-485) synthetic peptide whose sequence corresponds to the C-terminal of human ERb. The sequence of this peptide is (amino to carboxy terminus): C467-S-P-A-E-D-S-K-S-K-E-G-S-Q-N-P-Q-S-Q485This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and ERb , catalog &Num; PA1-313. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 mL of distilled water.Specifications
2100 | |
purified | |
Human Estrogen Receptor beta (aa 467-485) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q92731 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
ESR2 | |
No preservative | |
50 μg | |
C467 - S - P - A - E - D - S - K - S - K - E - G - S - Q - N - P - Q - S - Q485 |