missing translation for 'onlineSavingsMsg'
Learn More

Mouse Calreticulin (residues 24-43) Synthetic Peptide, Invitrogen™

Mouse Calreticulin (residues 24-43) Synthetic Peptide, PEP-018, from Invitrogen™

Brand:  Invitrogen PEP018

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11514680

  • £100.16 / 50µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-018 is a 20 amino acid (amino acids 24-43) synthetic peptide whose sequence is found in the amino-terminal of mouse calreticulin. The sequence of this peptide is (amino to carboxy terminus): K-E-Q-F-L-D-G-D-A-W-T-N-R-W-V-E-S-K-H-K-C (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native mouse calreticulin sequence) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and calreticulin, catalog # PA1-902A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 ml of distilled water.
Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
K-E -Q-F-L-D-G-D-A-W-T-N-R-W-V-E-S-K-H-K-C
Mouse Calreticulin (Residues 24-43) Synthetic Peptide
Videos