missing translation for 'onlineSavingsMsg'
Learn More

Mouse Calreticulin (residues 399-414) Synthetic Peptide, Invitrogen™

Mouse Calreticulin (residues 399-414) Synthetic Peptide, PEP-017, from Invitrogen™

Brand:  Invitrogen PEP017

UNSPSC : 12352203

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11504680

  • £97.00 / 50µg


Estimated Shipment
16-06-2017
Add to basket

Specifications

Description


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-017 is a 16 amino acid (amino acids 399-414) synthetic peptide whose sequence is found in the carboxy-terminal of mouse calreticulin. The sequence of this peptide is (amino to carboxy terminus): C-D-E-K-E-E-D-E-E-E-S-P-G-Q-A-K-D (The N-terminal cysteine has been added to facilitate conjugation and is not a part of the native mouse calreticulin sequence) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and calreticulin, catalog # PA1-903. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 ml of distilled water.
Description & Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
C-D-E-K-E-E-D-E-E-E-S-P-G-Q-A-K-D
Mouse Calreticulin (Residues 399-414) Synthetic Peptide