missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VAMP3 (aa 1-18), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Brand: Invitrogen PEP145
Code : LE
Additional Details : Weight : 0.01000kg
Description
Mouse VAMP-3 (residues 1-18) Synthetic Peptide, PEP-145, from Invitrogen.
PEP-145 is a 18 amino acid synthetic peptide whose sequence is from mouse VAMP-3. The sequence of this peptide is (amino to carboxy terminus): M (1)-S-T-G-V-P-S-G-S-S-A-A-T-G-S-N-R-R (18)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and the mouse VAMP-3, catalog # PA1-767A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
22319 | |
purified | |
Mouse VAMP3 (aa 1-18) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
P63024 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Vamp3 | |
No preservative | |
50 μg | |
M(1)-S-T-G-V-P-S-G-S-S-A-A-T-G-S-N-R-R(18) |