missing translation for 'onlineSavingsMsg'
Learn More

Bovine Prion Protein, (residues 146-159) Synthetic Peptide, Invitrogen™

Bovine Prion Protein (residues 146-159) Synthetic Peptide, PEP-051, from Invitrogen™

Brand:  Invitrogen PEP051

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11534710

  • £100.16 / 50µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-051 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 146-159 of bovine Prion Protein. The sequence of this peptide is (amino to carboxy terminus): S146-R-P-L-I-H-F-G-S-D-Y-E-D-R159 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and prion protein, catalog # MA1-750. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.
Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
S146-R-P-L-I-H-F-G-S-D-Y-E-D-R159
Bovine Prion Protein (Residues 146-159) Synthetic Peptide
Videos