Learn More
Invitrogen™ Human Myoglobin Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP75665
Code : LD
Weitere Details : CAS Number : 69-74-9 Weight : 0.01000kg
Beschreibung
Myoglobin (human) Recombinant Protein, RP-75665, from Invitrogen.
RP-75665 contains purified recombinant human Myoglobin protein with an N-terminal hexahistidine tag. RP-75665 may be used as a positive control in many immunological assays.Spezifikation
P02144 | |
20mM tris HCl with 250mM NaCl, 50% glycerol and no preservative; pH 8 | |
Human Myoglobin | |
10μg | |
N/A | |
RUO | |
MB | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control, Western Blot | |
4151 | |
8 | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human Myoglobin | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only