missing translation for 'onlineSavingsMsg'
Learn More

Luteinizing Hormone Releasing Hormone Synthetic Peptide, Invitrogen™

Luteinizing Hormone Releasing Hormone Synthetic Peptide, PEP-10934, from Invitrogen™

Brand:  Invitrogen PEP10934

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11564760

  • £478.17 / 25mg


Estimated Shipment
20-03-2018
Add to basket

Description

Description


Formulation: 25mg purified peptide and no preservative
PEP-10934 is a synthetic peptide suitable for use as a control in ELISA and Western blot applications. PEP-10934 has the amino acid sequence: Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2.
Specifications

Specifications

25mg
Purified
Yes
Store at -80°C, Avoid Freeze/Thaw Cycles
Luteinizing Hormone Releasing Hormone Synthetic Peptide
Control
25mg purified peptide and no preservative
Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2
LHRH, GnRH, Lutropin
Videos