Learn More
Invitrogen™ Rat FGF-7 (KGF) Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP10932
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
KGF (rat) Recombinant Protein, RP-10932, from Invitrogen.
The RP-10932 protein is a recombinant protein suitable for use as a control in ELISA and Western blot applications. RP-10932 is reported to have the additional biological activity: Binds to keratinocyts and most parenchymal epithelial cells; binds specific splice variant of FGF receptor expressed in epithelial cells (FGFr2IIIb).Specifications
Q02195 | |
PBS with no preservative | |
Rat KGF | |
E. coli | |
-20°C, Avoid Freeze/Thaw Cycles | |
Rat rKeratinocyte Growth Factor | |
Fgf7 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control | |
29348 | |
20μg | |
N/A | |
RUO | |
Rat KGF | |
Rat | |
Rt | |
Recombinant | |
Recombinant |
in vitro research only.