missing translation for 'onlineSavingsMsg'
Learn More

Interferon gamma (human) Recombinant Protein, Invitrogen™

Interferon gamma (human) Recombinant Protein, RP-75629, from Invitrogen™

Brand:  Invitrogen RP75629

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11558951

  • £155.76 / 10µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 10μg purified protein (0.23mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0
RP-75629 contains purified recombinant human IFN gamma protein with an N-terminal hexahistidine tag. RP-75629 may be used as a positive control in many immunological assays.
Specifications

Specifications

0.1mg/mL
Purified
Human Interferon gamma Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Control, Western Blot
10μg purified protein (0.23mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0
10μg
N/A
Unlabeled
E. coli
Videos