missing translation for 'onlineSavingsMsg'
Learn More

IL-8 (human) Recombinant Protein, Invitrogen™

IL-8 (human) Recombinant Protein, RP-75609, from Invitrogen™

Brand:  Invitrogen RP75609

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11588931

  • £429.24 / 100µg


Estimated Shipment
20-02-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein in TBS with 50% glycerol and no preservative; pH 8
RP-75609 contains a purified 77 amino acid fragment (aa23-99) corresponding to the mature human IL-8 protein with an N-terminal hexahistidine tag. RP-75609 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein in TBS with 50% glycerol and no preservative; pH 8
8
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Purified
Human IL-8 Recombinant Protein
100μg
N/A
Unlabeled
E. coli
Videos