missing translation for 'onlineSavingsMsg'
Learn More

IL-3 (human) Recombinant Protein, Invitrogen™

IL-3 (human) Recombinant Protein, RP-75593, from Invitrogen™

Brand:  Invitrogen RP75593

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11528921

  • £154.09 / 10µg


Estimated Shipment
21-02-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 10μg purified protein in TBS with 50% glycerol and no preservative; pH 8
RP-75593 contains purified recombinant human IL-3 protein with an N-terminal hexahistidine tag. RP-75593 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
10μg purified protein in TBS with 50% glycerol and no preservative; pH 8
8
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Purified
Human IL-3 Recombinant Protein
10μg
N/A
Unlabeled
E. coli
Videos