missing translation for 'onlineSavingsMsg'
Learn More

IL-3 (human) Recombinant Protein, Invitrogen™

IL-3 (human) Recombinant Protein, RP-75594, from Invitrogen™

Brand:  Invitrogen RP75594

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11538921

  • £520.32 / 100µg


Estimated Shipment
27-06-2018
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein in TBS with 50% glycerol and no preservative
RP-75594 contains purified recombinant human IL-3 protein with an N-terminal hexahistidine tag. RP-75594 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein in TBS with 50% glycerol and no preservative
100μg
N/A
Unlabeled
E. coli
Purified
Human IL-3 Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Videos