missing translation for 'onlineSavingsMsg'
Learn More

IL-12 p40 (human) Recombinant Protein, Invitrogen™

IL-12 p40 (human) Recombinant Protein, RP-75618, from Invitrogen™

Brand:  Invitrogen RP75618

UNSPSC : 12352203

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11578941

  • £426.00 / 100µg


Estimated Shipment
22-09-2017
Add to basket

Description

Description


Host Species: E. Coli
Formulation: 100μg purified protein in PBS with 50% glycerol and no preservative
RP-75618 contains purified recombinant human IL-12 p40 protein with an N-terminal hexahistidine tag. RP-75618 may be used as a positive control in many immunological assays.
Specifications

Specifications

Control, Western Blot
100μg purified protein in PBS with 50% glycerol and no preservative
Human IL-12 p40 Recombinant Protein
E. coli
store at -20°C, Avoid Freeze/Thaw Cycles
Human
Yes
Purified
16.75
100μg
N/A
Unlabeled
E. coli