Learn More
Human HTRA1, Recombinant Protein, Invitrogen™
Recombinant Protein
Brand: Invitrogen RP77541
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
HtrA1 Recombinant Protein, RP-77541, from Invitrogen.
HtrA1 inhibits signaling by proteins of the TGF family; degrades fragments of amyloid precursor protein; and shows increased expression in osteoarthritic cartilage. Truncated, recombinant human HtrA1 is expressed in E.coli cells. The protein consists of amino acids 141-480 of full-length HtrA1, a C-terminal 6x His-tag, and contains the catalytic and PDZ domain. RP-77541 is a truncated, recombinant human HtrA1 protein expressed in E. coli; amino acids 141-480 of full-length HtrA1 with a C-terminal 6x His-tag,containing the catalytic and PDZ domain. RP-77541 may be used as a control with corresponding anti-HtrA1 antibodies or in an enzymatic assay. RP-77541 appears as a major protein of about 37 kDa in SDS-PAGE.Specifications
Q92743 | |
Control | |
5654 | |
Human HTRA1 | |
5μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Human HTRA1 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.2mg/mL | |
50mM sodium phosphate with 300mM NaCl and no preservative; pH 7.5 | |
∽37 | |
7.5 | |
E. coli | |
RUO | |
HTRA1 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
in vitro research only