missing translation for 'onlineSavingsMsg'
Learn More

HIV1 Reverse Transcriptase Recombinant Protein, Invitrogen™

HIV1 Reverse Transcriptase Recombinant Protein, RP-4913, from Invitrogen™

Brand:  Invitrogen RP4913

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11584191

  • £1687.18 / 1000 units


Estimated Shipment
20-07-2018
Add to basket

Description

Description

HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1 (AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
Host Species: E. Coli
Formulation: 1000 units purified protein (0.37mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
RP-4913 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 Reverse Transcriptase (PA1-4948, PA1-4949, or PA1-4950), as well as in screening of specific inhibitors for new AIDS treatment . RP-4913 is composed of two subunits with calculated molecular weights of ∽66 kDa and ∽51 kDa. RP-4913 has a specific activity of 10,000-20,000 units/mg . Purity is 90%.
Specifications

Specifications

Control, Western Blot
1000 units purified protein (0.37mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
HIV1 Reverse Transcriptase Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Purified
∽66 and ∽51
8.3
1000 UN
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Videos