missing translation for 'onlineSavingsMsg'
Learn More

HIV1 Reverse Transcriptase Recombinant Protein, Invitrogen™

HIV1 Reverse Transcriptase Recombinant Protein, RP-4912, from Invitrogen™

Brand:  Invitrogen RP4912

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11574191

  • £425.20 / 200 units


Estimated Shipment
15-02-2018
Add to basket

Description

Description

HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1 (AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
Host Species: E. Coli
Formulation: 200 units purified protein (4000 units/ml) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
RP-4912 can be used as a positive control in Western blotting with ABR antibodies againt -HIV-1 Reverse Transcriptase, as well as in screening of specific inhibitors for new AIDS treatment . RP-4912 is composed of two subunits with calculated molecular weights of ∽66 kDa and ∽51 kDa; see figure 1. RP-4912 has a specific activity of ∽4,000 units/ml
Specifications

Specifications

4000U/mL
Purified
∽66 and ∽51
8.3
200 UN
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Control, Western Blot
200 units purified protein (4000 units/ml) in TBS with 50% glycerol, 0.01M beta mercaptoethanol, 5mMmgCl2, 0.1% Triton X-100 and no preservative; pH 8.3
HIV1 Reverse Transcriptase Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Videos