missing translation for 'onlineSavingsMsg'
Learn More

HIV1 p55 Recombinant Protein, Invitrogen™

HIV1 p55 Recombinant Protein, RP-4921, from Invitrogen™

Brand:  Invitrogen RP4921

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11564201

  • £1324.84 / 100µg


Estimated Shipment
20-02-2018
Add to basket

Description

Description

HIV-1 Gag p55 is a precursor protein of several proteins that form the core structure of AIDS virus, indispensable to its reproduction. This protein is digested by HIV-1 protease, first into intermediate products p41 and p15. Then p41 is digested into matrix protein p17 and capsid protein p24. Protein p15 is further digested into nucleocapsid protein p7 and to p6 and p1, both of unknown function.
Host Species: E. Coli
Formulation: 100μg purified protein (0.44mg/mL) in TBS with 20% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4921 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p55 (PA1-4959, or PA1-4960), as well as a p55 antigen in detection of anti-HIV-1 p55 antibody in Western blotting or ELISA. Also, it can be used as standard for the quantitative analysis of HIV-1 p55 antigen. RP-4921 has a calculated molecular weight of ∽55 kDa. Purity is 90%.
Specifications

Specifications

0.5mg/mL
Purified
∽55
7.5
100μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Control, ELISA, Western Blot
100μg purified protein (0.44mg/mL) in TBS with 20% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
HIV1 p55 Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Videos