missing translation for 'onlineSavingsMsg'
Learn More

HIV1 p17 Recombinant Protein, Invitrogen™

HIV1 p17 Recombinant Protein, RP-4915, from Invitrogen™

Brand:  Invitrogen RP4915

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11504201

  • £963.83 / 100µg


Estimated Shipment
27-06-2018
Add to basket

Description

Description

HIV-1 Gag p17 is the matrix protein of AIDS virus HIV-1 and is processed by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is indispensable to the reproduction of AIDS virus and constitute the essential element of the of AIDS virus particle construction.
Host Species: E. Coli
Formulation: 100μg purified protein in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4915 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p17 (PA1-4954, PA1-4955, or PA1-4956), as well as a p17 antigen in detection of anti-HIV-1 p17 antibody in Western blotting or ELISA. Also, it can be used as standard for the quantitative analysis of HIV-1 p17 antigen. RP-4915 has a calculated molecular weight of ∽17 kDa. Purity is 90%.
Specifications

Specifications

0.48mg/mL
Purified
∽17
7.5
100μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Control, ELISA, Western Blot
100μg purified protein in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
HIV1 p17 Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Videos