missing translation for 'onlineSavingsMsg'
Learn More

HIV1 p15 Recombinant Protein, Invitrogen™

HIV1 p15 Recombinant Protein, RP-4919, from Invitrogen™

Brand:  Invitrogen RP4919

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11544201

  • £1290.69 / 100µg


Estimated Shipment
20-07-2018
Add to basket

Description

Description

HIV-1 Gag p15 is processed by the digestion of its precursor Gag p55 by HIV-1 protease. This protein is further digested into nucleocapsid protein p7 and into p6 and p1 of unknown function. This digestion is promoted by the binding of HIV-1 genome RNA and the two Zn finger motifs that exist in the p7 region. The produced nucleocapsid protein p7 regulates the RNA function by directly binding to HIV-1 genome RNA.
Host Species: E. Coli
Formulation: 100μg purified protein (0.42mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
RP-4919 can be used as a positive control in Western blotting with ABR antibody anti-HIV-1 p15 (PA1-4957, or PA1-4958), as well as a p15 antigen in detection of anti-HIV-1 p15 antibody in Western blotting or ELISA. Also, it can be used as standard for the quantitative analysis of HIV-1 p15 antigen. RP-4919 has a calculated molecular weight of ∽15 kDa. Purity is 90%.
Specifications

Specifications

0.48mg/mL
Purified
∽15
7.5
100μg
store at -20°C, Avoid Freeze/Thaw Cycles
E. coli
Control, ELISA, Western Blot
100μg purified protein (0.42mg/mL) in TBS with 50% glycerol, 0.01M beta mercaptoethanol and no preservative; pH 7.5
HIV1 p15 Recombinant Protein
Chromatography
E. coli
Unlabeled
Yes
Videos